Learn More
Lenz Laborglasintrumente Extractor Heads for Specific Light Solvents
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05432149
 View more versions of this product
Product Code. 10409752
  • £304.00 / Each
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For the extraction of liquids with specific light solvents
  • With insert filter (porosity: 0)
  • Made of DURAN™ tubing
Specifications
Specifications
250 mL
Fritte P0
NS 45/40