Learn More
Lenz Laborglasintrumente Extractors Acc. to Soxhlet with Allihn Condenser
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05365025
 View more versions of this product
Product Code. 10614012
  • £229.00 / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Extractor in compact and robust design
  • With flask connection
  • Made of DURAN™ tubing
  • Allihn condenser with thread GL 14
  • Round bottom flask
Specifications
Specifications
30 mL
Duran Glass
NS 29/32