Learn More
Lenz Laborglasintrumente Extractors Acc. to Twisselmann
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05380031
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10283951
  • £597.00 / Each
Estimated Shipment: 27-09-2024
to see stock.
Add to basket
Description
Description
  • Extractor head with vacuum jacket
  • Made of DURAN™ tubing
  • Twisselmann condenser (improved) with thread GL 14
  • Round bottom flask
Specifications
Specifications
70 mL
Duran Glass
NS 34/35