Learn More
Lenz Laborglasintrumente Extractors for Solids
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05431031
 View more versions of this product
Product Code. 10356121
  • £248.00 / Each
Estimated Shipment: 26-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Extractor head with glass thimbles for solids
  • Made of DURAN™ tubing
  • Dimroth condenser (details on page 71)
  • With thread GL 14
  • Round bottom flask
Specifications
Specifications
70 mL
Borosilicate Glass 3.3
NS 45/40