Learn More
Lenz Laborglasintrumente Extractors for Specific Light Solvents
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05432049
 View more versions of this product
Product Code. 10674582
  • £537.00 / Each
Estimated Shipment: 26-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Extractor head with insert filter (frit, porosity: 0)
  • With PTFE valve stopcock
  • Made of DURAN™ tubing
  • Dimroth condenser
  • With thread GL 14
  • Round bottom flask
Specifications
Specifications
500 mL
0
NS 45/40