Learn More
Micronova™ Extreme Temperature Polyimide Cleanroom Tape
Extreme temperature polyimide tape is suitable for high heat and sub zero applications (-100°F to 500°F). The pressure-sensitive silicone adhesive ensures a clean release from most substrates, even after extreme temperature exposure.
Brand:  Micronova™ 825-1AM1
 View more versions of this product
Product Code. 16584512
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The Micronova Extreme Temperature Polyimide Cleanroom Tape features transparent amber polyimide film with pressure-sensitive silicone adhesive. Designed with extreme temperature resistance, this tape is ideal for heat sealing, solder mask for circuit boards, flame resistant electrical insulation, lamination, cold storage, lyophilization and freeze drying.
  • Transparent amber polyimide film with pressure-sensitive silicone adhesive
  • Extreme temperature resistant properties: -100°F to 500°F (-70°C to 260°C)
  • Suitable for high heat and sub zero applications
  • Clean release from most substrates, even after extreme temperature exposure
  • Ideal for flame resistant electrical insulation, solder mask for circuit boards, heat sealing, lamination, cold storage, lyophilization and freeze drying
  • Resistant to most chemicals including hydrocarbons, alcohols, and halogenated solvents
  • Wound on a plastic core
Specifications
Specifications
Not Autoclavable
Silicone, Pressure-sensitive
Amber
Tape
Extreme Temperature Cleanroom Tape
Suitable for high heat and subzero applications and ensures a clean release on Ideal for flame resistant electrical insulation, solder mask for circuit boards, heat sealing, lamination and test applications, over-labeling, subzero storage, lyophilization and freeze drying
Yes
16°C to 27°C
22 yd.
1 in.
1 in.
Certificate of Conformance, ISO 9001 Certified
Polymer
High Gloss
Polyimide
Non-sterile
MicronovaTM P52W Tape Dispenser
60°F to 80°F
20 m x 25.4 mm (22 yd. x 1 in.)
20 m
25.4 mm
25.4 mm