missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EYA4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24691-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the eyes absent (EYA) family of proteins. The encoded protein may act as a transcriptional activator through its protein phosphatase activity, and it may be important for eye development, and for continued function of the mature organ of Corti. Mutations in this gene are associated with postlingual, progressive, autosomal domit hearing loss at the deafness, autosomal domit non-syndromic sensorineural 10 locus. The encoded protein is also a putative oncogene that mediates DNA repair, apoptosis, and innate immunity following DNA damage, cellular damage, and viral attack. Defects in this gene are also associated with dilated cardiomyopathy 1J. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Specifications
EYA4 | |
Polyclonal | |
Unconjugated | |
EYA4 | |
CMD1J, DFNA10, EYA4, Eyes absent homolog 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14051, 2070 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95677, Q9Z191 | |
EYA4 | |
EYA4 Fusion Protein Ag20466 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |