missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Eye wash cabinet, contains 2 x 500ml of eye wash
Brand: Manutan A351860
Code : Z3
Additional Details : Weight : 1.00000kg
Description
Quick release wall bracket fixing. Contains 2 x 500ml bottles of sterile eye wash solution and two sterile eye pads with bandage.Dimensions H x D : 300mm x 120mm