Learn More
Kartell Eye Wash Station (German Version)
Emergency eye wash station moulded in strong high-impact PS. The directions are clearly printed on the support rack and the bottle - to be bought separately - can be filled with distilled water or other medicated liquid for ophthalmic use.
Brand:  Kartell 0238700
Product Code. 18037821
  • £311.00 / Pack of 10
Estimated Shipment: 17-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Emergency eye wash station moulded in strong high-impact PS
  • The directions are clearly printed on the support rack and the bottle can be filled with distilled water or other medicated liquid for ophthalmic use
  • The head has an air-vent valve to assure air refill of free space in the bottle thus preventing draw back of contaminated wash into the bottle
  • A perforated spray plate in the cup centre breaks up the stream to give the eye a gentle wash
  • A drain tube releases waste
Specifications
Specifications
Green
Wall
Non-sterile
Polystyrene
Eye Wash Station