Learn More
Sterile Eye Wash Bottle
Keep a Sterile Eye Wash Bottle set on hand as a first aid kit refill per the Health and Safety ACOP L74 regulation for contingency supplies in case of emergency.
Brand:  Safety First E404PL1
Product Code. 12424940
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The 1997 Approved Code of Practice states that 'Where tap water is not readily available for eye irrigation, at least 1L of sterile water or sterile normal saline 0.9% in sealed disposable containers should be provided'.
Specifications
Specifications
Eyewash Bottle
Eyewash Station
500 mL
Contain a Sterile Solution of Sodium Chloride (0.9%)
Sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products