Learn More
Thermo Scientific™ EZ-Link™ BMCC-Biotin
Biotinylate reduced cysteines (-SH groups) with a maleimide-activated, sulfhydryl-reactive biotinylation reagent containing a stabilizing cyclohexane group in the spacer arm.
Brand: Thermo Scientific™ 21900
Code : PN
Additional Details : Weight : 1.81440kg
Description
Thermo Scientific™ EZ-Link BMCC-Biotin is a maleimide-activated, sulfhydryl-reactive biotinylation reagent with an extended spacer arm that contains a stabilizing cyclohexane group.
Highlights:
- Biotinylates at pH 6.5-7.5
- Reaction avoids potential modification of tyrosine groups that can inactivate biological activity
- Longer spacer arm than iodoacetyl biotin
- Must be dissolved in DMSO or DMF before use
Specifications
BMCC-Biotin | |
Store at 4°C. | |
Cell-Permeant | |
Maleimide | |
Biotin | |
Biotin & Analogs |
50 mg | |
EZ-Link™ | |
Thiol | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide) | |
Extra-long |
For Research Use Only. Not for use in diagnostic procedures.