missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ DBCO Protein Labeling Kit

Kit provides everything needed to covalently label proteins with a dibenzylcyclooctyne (DBCO) group for copper-free click chemistry applications. Contains all necessary materials to perform five labeling reactions and subsequent purification for removal of excess DBCO reagent.

Brand:  Thermo Scientific™ C20046

Product Code. 17737483

  • £357.00 / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Kit provides everything needed to covalently label proteins with a dibenzylcyclooctyne (DBCO) group for copper-free click chemistry applications
  • Contains all necessary materials to perform five labeling reactions and subsequent purification for removal of excess DBCO reagent
  • TFP-PEG4-DBCO is a heterobifunctional crosslinker which contains an amine reactive tetrafluorophenyl (TFP), and an azide reactive dibenzylcyclooctyne (DBCO) with a water solubilizing PEG4 spacer in between
  • It readily reacts with the primary amines found on proteins to form a covalently attached DBCO group, which can be further reacted with an azide labeled coupling partner (supplied separately) to create diverse bioconjugates
  • In the first conjugation step, the TFP group reacts with a primary amine on a protein to form a stable amide bond and a covalently bound DBCO moiety
  • Subsequent purification removes excess crosslinker from solution which may interfere with the second labeling step
  • Once a protein is DBCO labeled and purified, it can undergo a copper free strain-promoted azide-alkyne cycloaddition (SPAAC)
  • Label proteins (20 to 150 kDa) with concentrations ranging from 0.5 to 5 mL
  • TFP esters are more hydrolytically stable than NHS esters enabling less reagent to be used
  • Labeling and purification can be completed in as little as 90 minutes with 30 min hands-on time
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 Kit
Chemical Reactivity
Wet Ice
EZ-Link™
Contains 5 vials of TFP Ester-PEG4-DBCO, 5 Zeba Dye & Biotin Removal columns, anhydrous DMSO, and BupHTM Phosphate Buffered Saline pack.

Store TFP Ester-PEG4-DBCO desiccated at –20°C, Zeba Dye and Biotin Removal columns at 2–8°C.

Chemical Labeling
TFP Ester, DBCO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers