Learn More
Thermo Scientific™ EZ-Link™ Iodoacetyl-LC-Biotin
Biotinylate reduced cysteines (-SH groups) at high pH via irreversible thioether bonds with this haloacetyl-activated, sulfhydryl-reactive biotinylation reagent.
Brand: Thermo Scientific™ 21333
Code : PN
Additional Details : CAS Number : 9048-46-8 Weight : 0.80000kg
Description
Thermo Scientific™ EZ-Link Iodoacetyl-LC-Biotin is a mid-length, haloacetyl-activated, sulfhydryl-reactive biotinylation reagent that forms stable, irreversible thioether bonds at alkaline pH.
Highlights:
- Reacts with sulfhydryl groups at pH 7.5-8.5
- Must be dissolved in DMSO or DMF before use
- Forms a stable thioether bond
Specifications
Iodoacetyl-LC-Biotin | |
Store desiccated at 4°C. Shipped at ambient temperature. | |
Cell-Permeant | |
Haloacetyl | |
Biotin | |
Biotin & Analogs |
50 mg | |
EZ-Link™ | |
Thiol | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide) | |
Long |
For Research Use Only. Not for use in diagnostic procedures.