Learn More
Thermo Scientific™ EZ-Link™ Iodoacetyl-PEG2-Biotin
Biotinylate reduced cysteines (-SH groups) at high pH with this haloacetyl-activated, sulfhydryl-reactive biotinylation reagent containing polyethylene glycol for solubility.
Brand: Thermo Scientific™ 21334
Code : PN
Additional Details : Weight : 0.22680kg
Description
Thermo Scientific™ EZ-Link Iodoacetyl-PEG2-Biotin is a mid-length, haloacetyl-activated, sulfhydryl-reactive biotinylation reagent that contains a 2-unit ethylene glycol in its spacer arm for increased water-solubility characteristics.
Highlights:
- Water-soluble, polyethylene glycol (PEG) spacer arm
- Reacts with sulfhydryl (-SH) groups at basic to neutral pH
- Coupling reactions occur in the dark
- PEG spacer arm transfers hydrophilicity to final conjugate
- Forms a stable thioether bond
Specifications
Iodoacetyl-PEG2-Biotin | |
Store desiccated at 4°C. Shipped at ambient temperature. | |
Cell-Impermeant | |
Haloacetyl | |
Biotin | |
Biotin & Analogs |
50 mg | |
EZ-Link™ | |
Thiol | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water | |
Mid-length, Pegylated |
For Research Use Only. Not for use in diagnostic procedures.