Learn More
Thermo Scientific™ EZ-Link™ Maleimide-PEG11-Biotin
Biotinylate reduced cysteines with this long, maleimide-activated, sulfhydryl-reactive biotinylation reagent with an 11-unit PEG spacer arm to enhance solubility and reach.
Brand: Thermo Scientific™ 21911
333.45 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Thermo Scientific™ EZ-Link Maleimide-PEG11-Biotin is a long, maleimide-activated, sulfhydryl-reactive biotinylation reagent that includes an 11-unit polyethylene glycol spacer arm for increased water-solubility and reach.
Highlights:
- Polyethylene glycol (PEG) spacer yields biotin-labeled proteins with greater solubility and less aggregation
- Forms stable thioether bond at pH 6.5-7.5
- PEG spacer improves aqueous solubility, allowing labeling to be performed under non-denaturing, physiological conditions
- Extended spacer arm reduces steric hindrance in binding to avidin, streptavidin and Thermo Scientific NeutrAvidin Proteins

Specifications
Maleimide-PEG11-Biotin | |
Store desiccated at 4°C. Shipped at ambient temperature. | |
Cell-Impermeant | |
Maleimide | |
Biotin | |
Biotin & Analogs |
25 mg | |
EZ-Link™ | |
Thiol | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water | |
Extra-long, Pegylated |
Safety and Handling
- EZ-Link Maleimide-PEG11-Biotin
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1A
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H350-May cause cancer.
- H360D-May damage the unborn child.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid, nickel(II) salt
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.