missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ EZ-Link™ NHS-LC-Biotin
Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Thermo Scientific™ EZ-Link NHS-LC-Biotin is a long-chain, NHS-ester activated biotinylation reagent for labeling primary amines (e.g., protein lysines), whose membrane permeability enables it to be used for general intracellular labeling.
Highlights:
- Extended spacer arm reduces steric hindrance when binding several biotinylated molecules to one avidin complex
- Less water-soluble than Sulfo-NHS-LC-Biotin
- Dissolve in organic solvent (DMSO or DMF) before use
- Penetrates cell membranes because it is uncharged
Specifications
Specifications
| Product Type | NHS-LC-Biotin |
| Quantity | 50 mg |
| Content And Storage | Store desiccated at 4°C. Shipped at ambient temperature. |
| Product Line | EZ-Link |
| Cell Permeability | Cell-Permeant |
| Chemical Reactivity | Amine |
| Reactive Moiety | Active Ester, Succinimidyl Ester, NHS Ester |
| Solubility | DMF (Dimethylformamide), DMSO (Dimethylsulfoxide) |
| Label or Dye | Biotin |
| Spacer | Mid-length |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction