missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ NHS-LC-LC-Biotin

Biotinylate protein amines with this doubly-long-chain (30.5A), NHS-ester biotinylation reagent containing a membrane permeable spacer arm for intracellular labeling.

Brand:  Thermo Scientific™ 21343

Product Code. 11841215

  • £246.00 / 50mg
Estimated Shipment: 25-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ EZ-Link NHS-LC-LC-Biotin is a doubly long-chain, NHS-ester activated biotinylation reagent for labeling primary amines (e.g., protein lysines), whose membrane permeability enables it to be used for intracellular labeling.

Highlights:

  • Longest spacer arm available to reduce steric hindrance when binding several biotinylated molecules to one avidin complex
  • Less water-soluble than sulfonated form
  • Dissolve in organic solvent (DMSO or DMF) before use
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NHS-LC-LC-Biotin
Store desiccated at 4°C. Shipped at ambient temperature.
Cell-Permeant
Active Ester, Succinimidyl Ester, NHS Ester
Biotin
Biotin & Analogs
50 mg
EZ-Link™
Amine
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Long
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.