Learn More
Thermo Scientific™ EZ-Link™ NHS-PEG12-Biotin, No-Weigh™ Format
Biotinylate protein amines and maximize solubility of proteins with this very long (56.0A) NHS-ester biotinylation reagent containing a pegylated, water-soluble spacer arm.
£230.00 - £948.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15792029
|
Thermo Scientific™
A35389 |
10 x 1 mg |
£230.00
10mg |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
11801195
|
Thermo Scientific™
21312 |
25 mg |
£267.00
25mg |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
11811195
|
Thermo Scientific™
21313 |
500 mg |
£948.00
500mg |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
Description
Thermo Scientific™ EZ-Link NHS-PEG12-Biotin is very long, pegylated biotinylation reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.
Highlights:
- Water-soluble polyethylene glycol (PEG) spacer arm increases solubility of labeled protein
- Excellent purity, molecular weight uniformity and amine-reactivity
- Long spacer arm reduces steric hindrance when binding to avidin molecules
Specifications
Biotin & Analogs |
For Research Use Only. Not for use in diagnostic procedures.