Learn More
Thermo Scientific™ EZ-Link™ NHS-PEG4-Biotin
Thermo Scientific™ EZ-Link™ NHS-PEG4-Biotin is a pegylated, water-soluble reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.
Brand:  Thermo Scientific™ 21363
 View more versions of this product
Product Code. 10464005
  • £956.00 / 1g
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ EZ-Link NHS-PEG4-Biotin is a pegylated, water-soluble reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.
  • Protein labeling - biotinylate antibodies or other proteins for detection or purification using streptavidin probes or resins
  • Amine-reactive - reacts with primary amines (-NH2), such as the side-chain of lysines (K) or the amino-termini of polypeptides
  • Pegylated - spacer arm contains a hydrophilic, 4-unit, polyethylene glycol (PEG) group
  • Enhances solubility - pegylation imparts water solubility to the biotinylated molecule, helping to prevent aggregation of biotinylated antibodies stored in solution
  • Irreversible - forms permanent amide bonds; spacer arm cannot be cleaved
  • Long reach - spacer arm (total length added to target) is 29 angstroms; this reduces steric hindrance when binding to avidin molecules
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EZ-Link NHS-PEG4-Biotin
1 g
EZ-Link™
Amine
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Long, Pegylated
NHS-PEG4-Biotin
Store at -20°C.
Cell-Impermeant
Active Ester, Succinimidyl Ester, NHS Ester
Biotin
Biotin & Analogs