Learn More
Thermo Scientific™ EZ-Link™ NHS-PEG4-Biotin
Thermo Scientific™ EZ-Link™ NHS-PEG4-Biotin is a pegylated, water-soluble reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.
Brand: Thermo Scientific™ 21362
Code : PN
Additional Details : Weight : 2.72160kg
Description
Thermo Scientific™ EZ-Link NHS-PEG4-Biotin is a pegylated, water-soluble reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.
- Protein labeling - biotinylate antibodies or other proteins for detection or purification using streptavidin probes or resins
- Amine-reactive - reacts with primary amines (-NH2), such as the side-chain of lysines (K) or the amino-termini of polypeptides
- Pegylated - spacer arm contains a hydrophilic, 4-unit, polyethylene glycol (PEG) group
- Enhances solubility - pegylation imparts water solubility to the biotinylated molecule, helping to prevent aggregation of biotinylated antibodies stored in solution
- Irreversible - forms permanent amide bonds; spacer arm cannot be cleaved
- Long reach - spacer arm (total length added to target) is 29 angstroms; this reduces steric hindrance when binding to avidin molecules
Specifications
EZ-Link NHS-PEG4-Biotin | |
50 mg | |
EZ-Link™ | |
Amine | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water | |
Long, Pegylated |
NHS-PEG4-Biotin | |
Store at -20°C. | |
Cell-Impermeant | |
Active Ester, Succinimidyl Ester, NHS Ester | |
Biotin | |
Biotin & Analogs |
For Research Use Only. Not for use in diagnostic procedures.