Learn More
Thermo Scientific™ EZ-Link™ NHS-PEG4-Biotin
Thermo Scientific™ EZ-Link™ NHS-PEG4-Biotin is a pegylated, water-soluble reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.
Brand: Thermo Scientific™ A39259
237.65 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Description
Thermo Scientific™ EZ-Link NHS-PEG4-Biotin is a pegylated, water-soluble reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.
- Protein labeling - biotinylate antibodies or other proteins for detection or purification using streptavidin probes or resins
- Amine-reactive - reacts with primary amines (-NH2), such as the side-chain of lysines (K) or the amino-termini of polypeptides
- Pegylated - spacer arm contains a hydrophilic, 4-unit, polyethylene glycol (PEG) group
- Enhances solubility - pegylation imparts water solubility to the biotinylated molecule, helping to prevent aggregation of biotinylated antibodies stored in solution
- Irreversible - forms permanent amide bonds; spacer arm cannot be cleaved
- Long reach - spacer arm (total length added to target) is 29 angstroms; this reduces steric hindrance when binding to avidin molecules
Specifications
NHS-PEG4-Biotin | |
Store at -20°C. | |
Wet Ice | |
Cell-Impermeant | |
NHS Ester | |
Biotin |
10 x 2 mg | |
Chemical Labeling | |
EZ-Link™ | |
Amine | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water | |
Pegylated, 29.0 Å |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.