Learn More
Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-Biotin
Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-Biotin is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.
Brand: Thermo Scientific™ 21335
308.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Code : PN
Additional Details : CAS Number : 127062-22-0 Weight : 1.81440kg
Description
Thermo Scientific™ EZ-Link Sulfo-NHS-LC-Biotin is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.
- Protein labeling - biotinylate antibodies to facilitate immobilization, purification or detection using streptavidin resins or probes
- Cell surface labeling - biotinylates only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
- Amine-reactive - reacts with primary amines (-NH2), such as lysine side-chains, or the amino-termini of polypeptides
- Soluble - charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
- Irreversible - forms permanent amide bonds; spacer arm cannot be cleaved
- Medium length - spacer arm (total length added to target) is 22.4 angstroms; provides excellent balance between adequate length (minimal steric hindrance for biotin binding) and modest mass
Specifications
EZ-Link Sulfo-NHS-LC-Biotin | |
100 mg | |
EZ-Link™ | |
Amine | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water | |
Mid-length |
Sulfo-NHS-LC-Biotin | |
Store desiccated at -20°C. Shipped at ambient temperature. | |
Cell-Impermeant | |
Active Ester, Succinimidyl Ester, Sulfo-NHS Ester | |
Biotin | |
Biotin & Analogs |
For Research Use Only. Not for use in diagnostic procedures.