missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ TFP Ester-PEG12-DBCO

Product Code. 17601586 Shop All Thermo Scientific Products
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 17601586

Brand: Thermo Scientific™ C20040

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A heterobifunctional crosslinker that contains an amine reactive tetrafluorophenyl (TFP) and a dibenzylcyclooctyne (DBCO) with a water solubilizing long PEG12 spacer in between. Use to label any primary amine containing molecule such as a protein to biomolecule with a distantly attached DBCO group.

  • A heterobifunctional crosslinker that contains an amine reactive tetrafluorophenyl (TFP) and a dibenzylcyclooctyne (DBCO) with a water solubilizing long PEG12 spacer in between the two groups
  • This crosslinker can be used to label any primary amine containing molecule such as a protein to biomolecule with a distantly attached DBCO group
  • Once a protein or (bio)molecule is DBCO labeled, it can undergo a copper-free azide-alkyne cycloaddition reaction to form a stable triazole linkage with an azide labeled coupling partner
  • Can be used to create diverse conjugates with the biorthogonal coupling between an alkyne and azide (supplied separately)
  • The coupling or crosslinking procedure is typically done in two steps:
    • First, a protein or amine containing molecule is labeled with a DBCO group and excess crosslinker is removed
    • Second, an azide containing moiety (prepared independently) is introduced into the same system as the DBCO moiety
    • Labeling occurs under biological conditions without the introduction of any additional reagents such as a copper catalyst
  • Reagent readily dissolves in water-miscible solvents for dilution into aqueous reaction mixtures
  • TFP esters are more hydrolytically stable than NHS esters
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Azide
Cleavable No
Description TFP Ester-PEG12-DBCO
Molecular Weight (g/mol) 1067.12
PEGylated Yes
Spacer Arm Length 46.3 Å
Content And Storage Upon receipt store desiccated at -20°C.
Cell Permeability Not determined
Shipping Condition Wet Ice
Product Line EZ-Link
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety TFP Ester, DBCO
Spacer Long (>30 Å)
Form Liquid
Quantity 25 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.