missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ezDNase™ Enzyme
SuperScript™ IV VILO™ Master Mix is a reaction master mix designed for fast, sensitive, and reproducible cDNA synthesis in RT-qPCR applications.
Brand: Invitrogen™ 11766051
111.72 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
SuperScript™ IV VILO™ (SSIV VILO) Master Mix is a reaction master mix designed for fast, sensitive, and reproducible cDNA synthesis in RT-qPCR applications. Inclusion of ezDNase, further accelerates the RT-qPCR workflow through an extremely simplified genomic DNA removal step. This master mix is also available without ezDNase, which can be purchased separately.Features of SuperScript IV VILO Master Mix include:
- Super-fast RT reaction in just 10 minutes and gDNA removal in 2 minutes
- Super-high yields—lower Ct values by more than 2 cycles ahead of all other reverse transcription reagents
- Super-convenient one-tube cDNA reaction master mix for two-step RT-qPCR
- Super-efficient reaction even with low template amounts and suboptimal purity samples
For even better performance, some SuperScript IV VILO Master Mix products include ezDNase (also available as a separate purchase) for genomic DNA removal to further accelerate the RT-qPCR workflow. The extremely simplified genomic DNA removal step dramatically reduces the time of the entire reverse transcription protocol and reduces possible variation in gene expression due to RNA loss or damage during conventional DNase treatment.
SSIV VILO master mix gives you more cDNA in less time with less pipeting, less variation, less reaction inhibition, less gDNA interference, and with less sample than all the other cDNA synthesis kits or master mixes for RT-qPCR.
Source
Purified from E. coli expressing the pol gene of M-MLV, modified to increase thermostability and half-life, processivity, inhibitor resistance, and to reduce RNase H activity.
Performance and quality testing
Assayed for endodeoxyribonuclease, exodeoxyribonuclease, and ribonuclease, as well as yield and length of cDNA product.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
10X | |
• ezDNase Enzyme, 50 μL • ezDNase Buffer (10X), 100 μL • Nuclease-free water, 1.25 mL Store at -5 to -30°C. |
|
DNase | |
50 μL | |
ezDNase™ | |
Liquid |
RT-qPCR | |
On dry ice | |
Reverse Transcription Reagents | |
Enzyme | |
RNA |
For Research Use Only. Not for use in diagnostic procedures.