Learn More
Invitrogen™ eBioscience™ eZKine™ 4-Color Compensation Control Kit
Designed for compensation setup with eZKine Whole Blood Intracellular Cytokine Kits. Contains necessary components to create single-color compensation controls for use with eZKine cocktails, including OneComp eBeads and 4 vials of antibody representing the 4 fluorochromes used in eZKine cocktails.
Brand: Invitrogen™ 8822-6856-41
Code : NEW
Additional Details : Weight : 0.23750kg
Alert:
Contains Sodium azide
Includes:
- OneComp eBeads: 1 drop/test, 100 tests
- Anti-Human CD3 (SK7) APC: 25 tests
- Anti-Human CD4 (SK3) PerCP-eFluor 710: 25 tests
- Anti-Human IL-17A (eBio64DEC17) PE: 25 tests
- Anti-Human IFN gamma (4S.B3) FITC: 25 tests
Description
OneComp eBeads can be stained with each of the 4 fluorochrome-conjuated antibodies to set up single-color compensation control tubes.Specifications
Human | |
CD3, CD4, IFN gamma, IL-17A | |
Flow Cytometric Analysis | |
2°C to 8°C. Do not freeze. Light sensitive material. |