missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One F-Bottom 96-well Assay Microplates

Product Code. 10177050
Click to view available options
Color:
Black
Clear
White
Sterility:
Non-sterile
Sterile
Lid:
With Lid
Without Lid
Unit Size:
Pack of 100
Pack of 120
Pack of 32
Pack of 40
This item is not returnable. View return policy

Product Code. 10177050

Brand: Greiner Bio-One 655180

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Features a chimney-like arrangement of the wells that allows each well to stand on its own. Greiner Bio One™ F-Bottom 96-Well Assay Microplates are constructed of polystyrene, a highly clear polymer with excellent optical properties making it ideal for precise optical measurements. Polystyrene is also characterized by its ability to bind biomolecules, such as proteins and is suitable for work with cell cultures.

  • Footprint compatible with automated systems
  • Can be traced to production through defined LOT number system
  • Free of detectable endotoxins (0.003 EU/mL) and regularly tested using an FDA-approved kinetic turbidimetric LAL-test (Limulus Amoebocyte Assay)
  • Manufactured from raw materials tested for leachables
  • Free of detectable DNase, RNase and human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance DIN ISO 9001
Certified RNase-/D-Nase, human DNA free
Format Standard
Lid With Lid
Culture Area 34 mm2
Array 8 x 12
Packaging Individually Wrapped
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 25 to 34 μL
Sterility Sterile
Surface Treatment Tissue Culture
Total Volume (Metric) Well 392 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
Max. Heat Stability -20 to +60°C
Max. RCF 4800 x g: Swinging-Bucket Rotor
Quantity 100/Cs.
RNase Free Yes
Temperature Range (Metric) -20°C to +60°C
Well Color Clear
Height (Metric) 11.5 mm
Diameter (Metric) Well 5 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.