missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ F1-ClipTip™ Multichannel Pipettes
GREENER_CHOICE

Ensure a complete seal on every channel with minimal tip attachment and ejection force using these multichannel pipetters, which feature an innovative locking technology.

£673.52 - £788.25

Specifications

Certifications/Compliance The F1-ClipTip pipetting system is CE-marked in accordance with IVD Directive in Europe. Manufactured according to ISO 9001, ISO 14001 and ISO 13485.
Model F1-ClipTip™
Product Line F1-ClipTip
Type Pipette
View More Specs

Products 8
Product Code Brand Volume (Metric) Number of Channels Compatible Tips Price Quantity & Availability  
Product Code Brand Volume (Metric) Number of Channels Compatible Tips Price Quantity & Availability  
13226299
View Documents
Thermo Scientific™
4661110
1 to 10 μL 8 F1-ClipTip 20 N/A N/A N/A
13236299
GREENER_CHOICE
View Documents
Thermo Scientific™
4661120
5 to 50 μL 8 F1-ClipTip 50 N/A N/A N/A
13266299
GREENER_CHOICE
View Documents
Thermo Scientific™
4661130
10 to 100 μL 8 F1-ClipTip 200 N/A N/A N/A
13246299
View Documents
Thermo Scientific™
4661140
30 to 300 μL 8 F1-ClipTip 300 N/A N/A N/A
13256299
View Documents
Thermo Scientific™
4661150
1 to 10 μL 12 F1-ClipTip 20 N/A N/A N/A
13276299
GREENER_CHOICE
View Documents
Thermo Scientific™
4661160
5 to 50 μL 12 F1-ClipTip 50 N/A N/A N/A
13286299
GREENER_CHOICE
View Documents
Thermo Scientific™
4661170
10 to 100 μL 12 F1-ClipTip 200 N/A N/A N/A
13206309
GREENER_CHOICE
View Documents
Thermo Scientific™
4661180
30 to 300 μL 12 F1-ClipTip 300 N/A N/A N/A
Description
Show More
Specifications

Specifications

The F1-ClipTip pipetting system is CE-marked in accordance with IVD Directive in Europe. Manufactured according to ISO 9001, ISO 14001 and ISO 13485.
F1-ClipTip™
F1-ClipTip
Pipette
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ F1-ClipTip™ Multichannel Pipettes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.