Learn More
Thermo Scientific™ F1-ClipTip™ Variable Volume Single Channel Pipettes
Lock tips firmly to the pipette with a complete seal with ClipTip technology, so they will not loosen or fall off. Lightweight handle design combined with minimal tip attachment and ejection forces.
Brand: Thermo Scientific™ 4641320N
203.81 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
Ensure a secure fit with ClipTip technology, which locks tips firmly to the pipette with a complete seal so they will not loosen or fall off. Lightweight handle design combined with minimal tip attachment and ejection forces will increase the conform in pipetting. Feel the difference.
Worry-free pipetting is finally here with the only tip that clips.Sealed in security of ClipTip Technology. Feel the difference.
Eliminate the frustration of banging tips on your pipette
With a light touch, each tip is clipped securely on the pipette.
Reduce the risk of loose or leaking tips
When ClipTip pipette tips are on, they are sealed and will not loosen. The ClipTip pipetting system remains sealed until you decide to eject tips, even in the most comprehensive applications, including multiple tip touches and mixing cycles.
Enable consistent and reproducible pipetting from user to user
Higher-quality results and more efficient research.
Learn more >
Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of repetitive strain injuries
Specifications
ClipTip 12.5 Ext | |
Pink | |
F1-ClipTip™ | |
1 to 10 μL | |
Pink |
F1-ClipTip | |
1 | |
Variable Volume Pipette | |
0.02μL |