missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP2/I-FABP Antibody (9A9B7B3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Bio-Techne NBP1-51589
319.20 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
FABP2/I-FABP Monoclonal specifically detects FABP2/I-FABP in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
FABP2/I-FABP | |
Monoclonal | |
Unconjugated | |
FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, I-FABPMGC133132, Intestinal-type fatty acid-binding protein | |
Mouse | |
15 kDa | |
0.1 mL | |
Cholesterol Metabolism, Lipid and Metabolism | |
2169 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
9A9B7B3 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
FABP2 | |
Purified recombinant fragment of human FABP2 expressed in E. Coli. | |
Unpurified | |
RUO | |
Primary | |
Human, Mouse | |
Ascites |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FABP2/I-FABP Antibody (9A9B7B3), Novus Biologicals™ > 0.1mL; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction