Learn More
Dräger™ Face Mask R52850 Full Panorama Nova RA
The Full Panorama Nova RA is ideal for applications involving hazardous materials, independence of the ambient air and where protection of the respiratory tract and eyes is needed. Designed for use with Rd40 filter range with it's RA connection. Filters must be purchased separately.
Brand: Dräger™ R52850
This item is not returnable.
View return policy
Description
- Full facemask
- Stainless steel fittings
- Rd40 standard thread connector
- Mist-free visor
- EPDM mask body, polycarbonate visor
- Can be used at high and low temperatures
- Comfortable fit - suitable for most face shapes
- 180° wide angle lens and unrestricted field of vision
Specifications
EN 136 class 3 (CE mark), DIN 58610, NIOSH, AS/NZS (depending on variant) | |
Rd 40 round thread x 1/7 in. | |
Mask body: EPDM, Visor: Polycarbonate |
Black | |
Full Facepiece Respirator |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Dräger™ Face Mask R52850 Full Panorama Nova RA > Full Panorama Nova RA Face Mask
Spot an opportunity for improvement?Share a Content Correction