Learn More
BD Facs Clean Solution
Used as a cleaning solution in flow cytometry systems. BD Biosciences™ Facs Clean Solution is RUO (GMP) regulatory compliant.
Brand:  BD 340345
Product Code. 15875858
  • £127.00 / Each
Explore more special offers
Description
Description
  • Flow cytometry (Routinely Tested)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 L
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • BD FACS Clean Solution
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P321-Specific treatment (see on this label).
  • P362-Take off contaminated clothing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Hypochlorous acid, sodium