Learn More
Invitrogen™ Human Factor VIIa, Native Protein
Native Protein
Brand: Invitrogen™ RP43108
Code : LD
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Factor VIIa (human) Native Protein, RP-43108, from Invitrogen.
RP-43108 contains native human factor VIIa protein. RP-43108 may be used as a positive control in many immunological assays. The RP-43108 protein molecular weight is ∽50 kDa. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!Specifications
P08709 | |
Control | |
2155 | |
20μg | |
-20°C, Avoid Freeze/Thaw Cycles | |
F7a | |
F7 | |
Unconjugated | |
Human | |
Liquid | |
purified |
2.0 mg/mL | |
50%water with 50% glycerol and no preservative | |
Human Factor VIIa | |
Human | |
RUO | |
Human Factor VIIa | |
Human | |
Hu | |
Native | |
Native |