missing translation for 'onlineSavingsMsg'
Learn More

Factor VIII Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications

£154.00 - £446.00

Specifications

Antigen Factor VIII
Concentration 1.0 mg/mL
Dilution Western Blot 2 ug/mL, ELISA reported in scientific literature, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18787073
View Documents
Novus Biologicals
NB100-91761SS
0.025 mg
£154.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18727803
View Documents
Novus Biologicals
NB100-91761
0.1 mg
£446.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Factor VIII Polyclonal specifically detects Factor VIII in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Factor VIII
Western Blot 2 ug/mL, ELISA reported in scientific literature, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin 1:50 - 1:200
Polyclonal
Rabbit
Cardiovascular Biology
PBS with 0.05% Sodium Azide
AHF, Antihemophilic factor, coagulation factor VIII, procoagulant component, coagulation factor VIIIc, DXS1253E, F8Ccoagulation factor VIII, factor VIII F8B, FVIII, HEMAF8B, Procoagulant component
F8
IgG
Affinity Purified
267 kDa
1.0 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Rat
P00451
2157
A synthetic peptide made to a C-terminal portion of human Factor VIII (between amino acids 2100-2250) [UniProt P00451]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.