missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Factor XIa, Native Protein
Native Protein
Brand: Invitrogen™ RP43122
Code : LD
Additional Details : Weight : 0.01000kg
Description
Factor Xia (human) Native Protein, RP-43122, from Invitrogen.
RP-43122 contains native human Factor XIa protein. RP-43122 may be used as a positive control in many immunological assays. The RP-43122 protein molecular weight is ∽160 kDa. Activity: 322 U/mg as determined by Factor XI clotting assay (1 unit = Factor XI activity in 1 ml of normal plasma). The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!Specifications
P03951 | |
Control | |
2160 | |
50μg | |
-20°C | |
F11a | |
F11 | |
Unconjugated | |
Human | |
Liquid | |
purified |
2.5 mg/mL | |
50%water with 50% glycerol and no preservative | |
Human Factor XIa | |
Human | |
RUO | |
Human Factor XIa | |
Human | |
Hu | |
Native | |
Native |