missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Factory Calibration (3 Temperatures) for UF30
Shop All Memmert Products
Click to view available options
Unit Size:
Each
Description
- For three temperature values: 100°C, 160°C, and 220°C
Specifications
Specifications
| For Use With (Equipment) | UF30 Oven |
| Description | Measuring point chamber center |
| Calibration Type | Certificate for 3 temperatures |
| Product Type | Standard Works Calibration Certificate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction