missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FADD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | FADD |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16803814
|
Proteintech
14906-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16893804
|
Proteintech
14906-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is an adaptor molecule that interacts with various cell surface receptors and mediates cell apoptotic signals. Through its C-terminal death domain, this protein can be recruited by TNFRSF6/Fas-receptor, tumor necrosis factor receptor, TNFRSF25, and TNFSF10/TRAIL-receptor, and thus it participates in the death signaling initiated by these receptors. Interaction of this protein with the receptors unmasks the N-terminal effector domain of this protein, which allows it to recruit caspase-8, and thereby activate the cysteine protease cascade. Knockout studies in mice also suggest the importance of this protein in early T cell development.Specifications
FADD | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q13158, Q61160 | |
14082, 266610, 8772 | |
FADD Fusion Protein Ag6701 | |
Primary | |
-20°C | |
FADD |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FADD, GIG3, MORT1, Protein FADD | |
FADD | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |