Learn More
3M™ Fahrenheit™ Anti-Scratch Safety Goggles
64.20 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Use promo code "22912" to get your promotional price.
Description
- Soft, crystal-clear PVC frame goggles with a wide support area all around the face to enhance comfort.
- Specific channel on the side of the frame to accommodate the temple of prescription frames.
- Model fits well with prescription glasses.
- Nylon elastic strap which is easily adjustable thanks to a buckle system and to a pivoting strap attachment mechanism.
- Indirect ventilation system to protect against liquid droplets and large dust particles.
- Anti-Scratch coating for increased durability.
Specifications
Certifications/Compliance | CE Approved , EN 166: 2001 |
Product Type | Safety Goggles |
For Use With (Application) | Construction, food industry |
Frame Color | Clear |
Ventilation Details | Indirect Vented |
Material | PVC |
Lens Material | Polycarbonate |
Frame Type | Soft Frame |