missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ 384-Well, Cell Culture-Treated, Flat-Bottom Microplate

Product Code. 11503520 Shop All Corning Products
Click to view available options
Color:
Clear
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 11503520

Brand: Falcon™ 353961

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Color:Clear; Sterility:Sterile; Lid:With Lid

  • Ideal for use in high throughput systems
  • Falcon 384-well microplates are compatible with automation systems and meet standard ANSI/SBS footprint dimensions for microplates
  • TRUSTED_SUSTAINABILITY

    Specifications

    Format Standard
    Lid With Lid
    Content And Storage Ambient
    Culture Area 10 mm2
    Array 16 x 24
    Binding Type TC-Treated
    Color Clear
    For Use With (Equipment) Automation systems
    Material Polystyrene
    No. of Wells 384
    Volume (Metric) Working 15 to 110 μL
    Sterility Sterile
    Surface Treatment Cell Culture
    Well Shape Flat Bottom (F-well)
    No. per Case 50/Cs.
    No. per Pack 5/Pk.
    Quantity 50/Cs.
    Well Color Clear
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.