Learn More
Falcon™ Cell Culture Multi Flasks
Grow cells faster and easier, increasing productivity in cell culture workflow. Falcon™ Cell Culture Mini Flasks provide even distribution of media across all layers for homogeneous cell growth.
£147.19 - £169.00
Specifications
Material | Polystyrene |
---|---|
Neck Style | Straight |
Shape | Rectangular |
Sterility | Sterile |
Closure Type | Vented |
Product Code | Brand | Culture Area | Surface Coating | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Culture Area | Surface Coating | Price | Quantity & Availability | ||||
11587421
|
Falcon™
353143 |
525 cm2 | 3 Layer TC-Treated |
£147.19
Pack of 12 |
In Stock
Log in to see stock. |
||||
11597421
|
Falcon™
353144 |
875 cm2 | TC-Treated |
£169.00
Pack of 8 |
In Stock
Log in to see stock. |
||||
Description
Increase productivity by enabling to grow more cells faster and easier. 3- or 5-layer formats provide 525 cm2 and 875 cm2 cell growth surface area, respectively.
- Even distribution of media across all layers for homogeneous cell growth
- Ability to mix cells and reagents in the Falcon Multi-Flask saves time
- Flexible design allows to pour or aspirate/recover cells using a pipet
- Consistent surface treatment for predictable scale-up
- Lot number printed on each flask for traceability
- Manufactured in compliance to cGMP standards
Specifications
Polystyrene | |
Rectangular | |
Vented |
Straight | |
Sterile |
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.