Learn More
Falcon™ Clear Assay Plate Lid
For 384-well (PS) and polyvinyl chloride (PVC) plates
Brand: Falcon™ 353958
Code : 66
Additional Details : Weight : 1.15000kg
Description
- Polystyrene lid for 384 well and 96 well Falcon™ microplates with 120μL total well capacity
- 6mm polystyrene, for 96 and 384 well microplates
- Non-sterile
Specifications
Polystyrene | |
Non-sterile | |
Microplate lid | |
96/384-well Plate, 120 μL Wells |