missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ Chambered Cell Culture Slides
BIOPHARMA

Packaged in disposable incubation trays. Falcon™ Chambered Cell Culture Slides are ideal for in situ DNA amplification/hybridization, tissue and tumor typing/pathology, or any study requiring a small, accessible culture area.

Brand:  Falcon™ 354104

 View more versions of this product

Product Code. 10693472

  • £627.00 / Pack of 96
Estimated Shipment: 29-05-2025
to see stock.
Explore more special offers
Add to basket

Includes: Handled polystyrene lid, safety removal tool
This item is not returnable. View return policy

Description

Description

 

  • Simplify analysis: culture, fix, stain and  examine your samples under a microscope without cell transfer
  • Polystyrene chamber affixed to a specially cleaned and triple-rinsed glass microscope slide (25 × 75mm)
  • Safety Removal Tool separates wells from slide; gasket remains with plastic vessel, prevents cross-contamination and doesn't interfere with further processing
  • Sterile
  • Culture up to 8 different conditions on a single glass microscope slide
  • Numeric labeling; 4-well, and 8-well formats available

Order Info

Available with 1, 2, 4, or 8 wells, numbered for easy identification. Supplied with handled polystyrene lid. Packaged in disposable incubation trays. Each tray holds 12 slides.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cell Culture Slide
1.7 cm2
In situ DNA amplification or hybridization, tissue and tumor typing or pathology, or any study requiring a small, accessible culture area
96
4
1 to 1.5 mL
Handled polystyrene lid, safety removal tool
Sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.