Learn More
Falcon™ HTS 24-Well Multiwell Permeable Support System
Save time and labor in HTS protocols—grow, handle, and analyze 24 samples simultaneously. Falcon™ HTS Multiwell Insert System features a generous, automation-compatible sampling port which allows sampling above and below membranes.
£118.77 - £999.16
Specifications
Color | Clear |
---|---|
Culture Area | 0.3 cm2 |
Sterility | Sterile |
Type | Multi-well Insert System |
Well Count | 24 |
Includes: 24-well insert plate, 24-well plate or single-well feeder tray, and lid
Product Code | Brand | Pore Size | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Quantity | Price | Quantity & Availability | ||||
10182511
|
Falcon™
351185 |
8 μm | 5/Cs. |
£552.37
Pack of 5 |
Estimated Shipment: 10-12-2024
Log in to see stock. |
||||
10326011
|
Falcon™
351183 |
3 μm | 5/Cs. |
£999.16
Pack of 5 |
Estimated Shipment: 10-12-2024
Log in to see stock. |
||||
10265141
|
Falcon™
351181 |
1 μm | 5/Cs. |
£558.50
Pack of 5 |
In Stock
Log in to see stock. |
||||
10693462
|
Falcon™
351184 |
8 μm | Each |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Falcon 24 Multiwell Insert Systems are designed to automate many commonly used cell-based assays for drug discovery. Available in a choice of membrane pore sizes, 24 Multiwell inserts have been successfully used for a variety of applications including permeability studies for oral bioavailability (e.g., Caco-2 cells), chemotaxis, cell migration and invasion assays. These insert systems offer all the benefits of Falcon Individual Cell Culture Inserts in an automation-friendly format that is compatible with most robots and fluid handlers. The Falcon 24 Multiwell Insert Plate is a single unit that is compatible with all Falcon 24 well plates and Feeder Tray.
Specifications
Clear | |
Sterile | |
24 | |
Polyethylene Terephthalate |
0.3 cm2 | |
Multi-well Insert System | |
Excellent for differentiation, toxicology, migration and invasion studies |