missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ 96-Well, Cell Culture-Treated, Flat-Bottom, Insert System Microplate

Color:Clear; Sterility:Sterile; Lid:With Lid

Brand:  Falcon™ 351130

Code : 66

Additional Details : Weight : 0.22000kg

Product Code. 10271102

  • £491.05 / Each
Estimated Shipment: 26-11-2024
to see stock.
Add to basket

Description

Description

The Falcon 96 Multiwell Insert System is a cell culture insert platform suitable for both manual and robotic screening of compounds in cell-based assays. The system has been tested for its ability to produce a differentiated monolayer of Caco-2, LLC-PK1, and MDCK cells making it an ideal platform for in vitro bioavailability and permeability studies.

This automation compatible platform is composed of a 1.0 μm pore size PET membrane-based 96 Multiwell Insert plate, a media feeder tray, and a lid. The drop-in baffle for the Feeder Tray mitigates media sloshing and lowers the risk of contamination. To analyze individual samples, simply transfer the insert plate into the Falcon 96 Square Well, Angled-Bottom Plate. If desired, the Falcon 96 Square Well, Angled-Bottom Plate may be used in conjunction with the insert for culturing the cells, eliminating the transfer step from the single-well feeder tray for sample analysis.

  • Automation compatible design
  • Format compatible with most robots and fluid handling instruments
  • Complete sample recovery
  • The Falcon 96 Square Well, Angled-Bottom Plate features an angled bottom for more complete sample utilization
  • Excellent reproducibility
  • One-piece feeder tray enhances consistency in well-to-well monolayer growth
  • Total assay flexibility—ideal for transport studies
  • System can be used with many cell lines including Caco-2, MDCK, and LLC-PK1, for basal to apical or apical to basal measurements of drug transport
Specifications

Specifications

Standard
0.08 cm2
8 x 12
Clear
Sterile
Flat Bottom (F-well)
Insert plate with feeder tray and lid
1/Pk.
With Lid
PET
1 μm
96
Cell Culture
To produce a differentiated monolayer of Caco-2 cells, suitable for bioavailability and permeability studies
1/Ea.
1/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers