Learn More
Falcon™ 96-Well, Cell Culture-Treated, Flat-Bottom, Insert System Microplate
Color:Clear; Sterility:Sterile; Lid:With Lid
Brand: Falcon™ 351131
Code : 67
Additional Details : Weight : 0.55000kg
Description
The Falcon 96 Multiwell Insert System is a cell culture insert platform suitable for both manual and robotic screening of compounds in cell-based assays. The system has been tested for its ability to produce a differentiated monolayer of Caco-2, LLC-PK1, and MDCK cells making it an ideal platform for in vitro bioavailability and permeability studies.
This automation compatible platform is composed of a 1.0 μm pore size PET membrane-based 96 Multiwell Insert plate, a media feeder tray, and a lid. The drop-in baffle for the Feeder Tray mitigates media sloshing and lowers the risk of contamination. To analyze individual samples, simply transfer the insert plate into the Falcon 96 Square Well, Angled-Bottom Plate. If desired, the Falcon 96 Square Well, Angled-Bottom Plate may be used in conjunction with the insert for culturing the cells, eliminating the transfer step from the single-well feeder tray for sample analysis.
- Automation compatible design
- Format compatible with most robots and fluid handling instruments
- Complete sample recovery
- The Falcon 96 Square Well, Angled-Bottom Plate features an angled bottom for more complete sample utilization
- Excellent reproducibility
- One-piece feeder tray enhances consistency in well-to-well monolayer growth
- Total assay flexibility—ideal for transport studies
- System can be used with many cell lines including Caco-2, MDCK, and LLC-PK1, for basal to apical or apical to basal measurements of drug transport
Specifications
Standard | |
0.08 cm2 | |
8 x 12 | |
Clear | |
Sterile | |
Flat Bottom (F-well) | |
5/Cs. | |
5/Cs. |
With Lid | |
PET | |
1 μm | |
96 | |
Cell Culture | |
Insert plate with feeder tray and lid | |
5/Pk. |