missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ Sterile Polyethylene Snap Caps
GREENER_CHOICEBIOPHARMA

Product Code. 10011021 Shop All Corning Products
Click to view available options
Color:
Natural
Unit Size:
Pack of 2000

Product Code. 10011021

Brand: Falcon™ 352032

Only null left
Add to basket
Edge
Add to basket

For Falcon Round-Bottom Tubes

These polyethylene caps feature a patented, two-position fit; loose for aerobic culturing, tight for sealing.

Falcon™ polyethylene dual-position snap caps for 12x75mm tubes offer both vented and fully closed positions. These sterile caps are packaged 500 per bag and 2,000 per case. RNase/DNase-free.
  • Dual position snap cap, offering vented as well as fully closed options
  • Sterilized by gamma irradiation
  • For use with both polystyrene and polypropylene tubes
  • Order Info

    Supplied in bags of 500; case of four bags.

    TRUSTED_SUSTAINABILITY

    Specifications

    Product Type Snap Cap
    Material Polyethylene
    Color Natural
    No. per Pack 500/Pk.
    No. per Case 2000/Cs.
    For Use With (Equipment) Falcon™ 75 L x 12 mm O.D. 5 mL polystyrene and polypropylene round bottom tubes
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.