missing translation for 'onlineSavingsMsg'
Learn More

Riedel-de Haën FAM Benzine

Product Code. 15915610
Click to view available options
Quantity:
18 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

FAM Benzine, DIN 51635, bp 65°C to 95°C.

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals.
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Specifications

pH Not applicable
Melting Point -30°C
CAS 110-82-7
CAS Min % ≥10.00%
Color Colorless
CAS Max % ≤25.00%
Density 0.76 g/cm3 (15°C)
Boiling Point 65°C to 95°C
Quantity 18 L
Flash Point -26°C
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Molecular Formula C6H12
MDL Number MFCD00003814
Vapor Pressure 10 hPa (20°C)
UN Number UN3295
Viscosity No data available
Synonym Petroleum ether, FAM-Petroleum ether, Petroleum benzin
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
SMILES C1CCCCC1
IUPAC Name cyclohexane
Molecular Weight (g/mol) 84.16
Grade ACS
Chemical Name or Material FAM Benzine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'