missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fam Benzine, Honeywell
Click to view available options
Quantity:
2.5 L
25 L
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 0.690 to 0.705g/mL at 15°C |
Flash Point | −26°C (−14.8°F) |
Quantity | 2.5 L |
UN Number | 1268 |
Chemical Name or Material | Fam Benzine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'