Learn More
FAM114A1 Mouse anti-Human, Unconjugated, Clone: 1B11D6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67926-1-IG-150UL
This item is not returnable.
View return policy
Description
The FAM114A1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human FAM114A1. This antibody recognizes human, rat, mouse antigen. The FAM114A1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
FAM114A1 | |
Monoclonal | |
Unconjugated | |
150 μL | |
92689 | |
IgG2a |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
1B11D6 | |
Mouse | |
Primary | |
Human, Mouse, Rat |