missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ FAM3™ Dye-Labeled Pre-miR Negative Control #1

Product Code. 10744997
Click to view available options
Quantity:
5 nmol
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10744997

Brand: Ambion™ AM17121

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

FAM3™ Dye-Labeled Pre-miR Negative Control #1

Ambion™ FAM™ dye-labeled synthetic miRNA mimic for miRNA functional analysis. 5 nmol is supplied.

  • Optimized design results in correct strand utilization by RISC
  • Purified and ready to transfect

    Ambion™ Pre-miR™ miRNA Precursor Molecules are small, chemically modified, double-stranded RNA molecules designed to mimic endogenous mature miRNAs. This ready-to-use miRNA mimic can be introduced into cells using transfection or electroporation parameters similar to those used for siRNAs. They enable detailed study of miRNA biological effects via gain-of-function experiments. The FAM™ Dye-Labeled Pre-miR™ Negative Control #1 is a random sequence Pre-miR™ molecule that has been extensively tested in human cell lines and tissues and validated to produce no identifiable effects on known miRNA function. It is ideal for monitoring transfection efficiency, as the fluorescent label enables direct observation of the cellular uptake, distribution, and localization of the control. The label is on the 5' end of the molecule, and the oligonucleotide sequence is the same as the Pre-miR™ miRNA Precursor Molecule Negative Control #1 (SKU #AM17110). NOTE: Pre-miR™ miRNA Precursors are not hairpin constructs and should not be confused with pre-miRNAs.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Pre-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. It is guaranteed for 1 year when stored dried at –20°C.
    Format Tube
    Control Type Transfection Control
    RNAi Type miRNA Inhibitor
    For Use With (Application) miRNA & Non-Coding RNA Analysis
    Label or Dye FAM
    Product Line Ambion, FAM, Pre-miR
    Product Type miRNA Mimic
    Purity HPLC
    Quantity 5 nmol
    Shipping Condition Dry Ice
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.