missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 7, Phosphate buffer, traceable to NIST, ready to use, Thermo Scientific Chemicals
£21.91 - £23.62
Specifications
Chemical Name or Material | Buffer solution pH 7 |
---|---|
Name Note | Phosphate buffer, traceable to NIST, ready to use |
CAS | 7558-79-4, 7732-18-5, 7487-94-7, 7778-77-0, 7647-14-5 |
Density | 1.0000g/mL |
MDL Number | MFCD03097944 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10706383
|
Thermo Scientific Acros
383855000 |
500 mL |
£21.91
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10026042
|
Thermo Scientific Acros
383850010 |
1 L |
£23.62
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer solution pH 7 | |
7558-79-4, 7732-18-5, 7487-94-7, 7778-77-0, 7647-14-5 | |
MFCD03097944 | |
Solubility in water: miscible. |
Phosphate buffer, traceable to NIST, ready to use | |
1.0000g/mL | |
1 |
Safety and Handling
Product Identifier
- Buffer solution pH 7, Phosphate buffer, traceable to NIST
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title